Order Entry
Czech Republic
ContactUsLinkComponent
Sigma-Aldrich® Zerostat anti-static instrument
Sigma-Aldrich® Zerostat anti-static instrument
Catalog # SIALZ108812-1EA
Supplier:  Merck
Sigma-Aldrich® Zerostat anti-static instrument
Catalog # SIALZ108812-1EA
Supplier:  Merck

Specifications

  • Description:
    Zerostat anti-static instrument, Zerostat 3
  • Pk:
    1

Specifications

About this item

The Zerostat 3 prevents charged particles from "flying" around during weighing or transfer, and keeps film, glass, and plasticware dust and lint-free.

  • With a unique ′Piezo Crystal′ device which neutralises static electricity
  • Portable and does not require batteries, electrical power supply or refills
  • Life expectancy is 50000+ squeeze cycles (handle action)
  • Each Zerostat 3 comes with a test ion-indicator to confirm that the unit is working properly

Material: ABS, acrylonitrile butadiene styrene
Size: 101,6×152,4 mm
Weight: 1,39 kg

Delivery: Zerostat 3 comes with a test ion-indicator.
Caution: Do not use in an explosive or combustible environments.