Order Entry
Czech Republic
ContactUsLinkComponent
Xylene (mixture of isomers) ≥98%, liquid
Flammable
Irritant
Xylene (mixture of isomers) ≥98%, liquid
Supplier:  PanReac AppliChem
Certificates
undefined
Flammable
Irritant
Xylene (mixture of isomers) ≥98%, liquid
Supplier:  PanReac AppliChem

Specification Test Results

  • Appearence:
    Clear colorless liquid
  • Solubility:
    Soluble in water 0,2 g/l at 20 °C
  • Vapor pressure:
    20 °C
  • Assay:
    98%
  • Refractive index n20/D:
    1,497
  • Ignition temperature:
    465 °C
  • Vapour pressure (20 °C) :
    8 hPa
  • Dielectric constant (25 °C):
    2,4
  • Evaporation number (DIN 53170):
    13,5
  • Saturation concentration (20 °C):
    35 g/m³
  • Explosion limit:
    1,7 − 7,6% (V)
  • Identity (IR):
    Passes test
  • Density at 20/4:
    0,862 – 0,864
  • Acidity:
    0,0003 meq/g
  • Alkalinity:
    0,00025 meq/g
  • Non-volatile matter:
    0,01%
  • Thiophene (C₄H₄S):
    0,0005%
  • Water (H₂O):
    0,05%
  • Copper (Cu):
    0,00002%
  • Iron (Fe):
    0,00005%
  • Nickel (Ni):
    0,00002%
  • Lead (Pb):
    0,00002%

Specification Test Results

Frequently Bought Together