Vydac 238MS C18, HPLC columns
Supplier: Avantor
Certificates
About this item
These Vydac MS columns are a later development from Grace for the reversed phase HPLC separation of biomolecules. A proprietary surface treatment and bonding process gives Vydac MS columns a unique selectivity. A variety of reversed phases makes this product line suitable for the analysis of small peptides to large intact, undenatured proteins.
- Five reversed-phase chemistries
- Excellent peak shape with little or no TFA
- High protein recoveries make scale-up easy
Monomeric bonding offers increased peptide interaction and generally yields higher peak counts. Use for same applications as 218MS, but offers different C18 selectivity.
Vydac is a registered trademark of W.R.Grace and Co.
Specifications
- Catalog No:
- 238GK54MS/N
- 238MS5.310
- Environmentally Preferable:
- Low Manufacturing Impact
- Carbon load:
- 4%
- Endcapped:
- Yes
- USP code:
- L1
- Environmental certification:
- ISO 14001:2015 (530840 UM15)