Rechargeable NiMH Battery
613-4252
: DRUMMOND
- Description:Rechargable NiMH batteries
- Pk:1
Replacement battery for Drummond Scientific Pipet-Aid® controllers.
NiMH 8.4v 200mAh for:
Portable Pipet-Aid® and Pipet-Aid® XP
3/VH 700AAA 3.6v 750mHh for:
Pipet-Aid® XP2 and Pipet-Aid® XL