Anti-TACI detection. Tested with WB, ELISA(Cap) in Human. Polyclonal Antibody
Catalog # BSBTA02952-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:TACI detection. Tested with WB, ELISA(Cap) in Human.
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for TACI detection. Tested with WB, ELISA(Cap) in Human.
Type: Primary
Antigen: TACI detection. Tested with WB, ELISA(Cap) in Human.
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human