Anti-MIF Polyclonal Antibody
Catalog # BSBTPB9274
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:MIF
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for MIF detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. MIF information: Molecular Weight: 12476 MW; Subcellular Localization: Secreted. Cytoplasm. Does not have a cleavable signal sequence and is secreted via a specialized, non- classical pathway. Secreted by macrophages upon stimulation by bacterial lipopolysaccharide (LPS), or by M.tuberculosis antigens.
Type: Primary
Antigen: MIF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human