Anti-Dynamin 2 Polyclonal Antibody
Catalog # BSBTA01629-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:Dynamin 2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Dynamin 2 detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: Dynamin 2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat