Specifications
- Antibody type:Primary
- Antigen symbol:ATP2A3/Serca3 Atpase
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for SERCA3 ATPASE/ATP2A3 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. SERCA3 ATPASE/ATP2A3 information: Molecular Weight: 113977 MW; Subcellular Localization: Nucleus membrane ; Multi-pass membrane protein . Endoplasmic reticulum membrane ; Multi-pass membrane protein . Sarcoplasmic reticulum membrane ; Multi-pass membrane protein ; Tissue Specificity: Found in most tissues. Most abundant in thymus, trachea, salivary gland, spleen, bone marrow, lymph node, peripheral leukocytes, pancreas and colon. Also detected in fetal tissues. Expressed in cell lineages of hematopoietic, epithelial, or embryonic origin and also expressed in several cancer cell lines.
Type: Primary
Antigen: ATP2A3/Serca3 Atpase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat