
Potassium dihydrogen phosphate, anhydrous, white crystalline powder cell culture reagent
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7778-77-0
:
Formula:KH₂PO₄
MDL Number:MFCD00011401
:136.09 g/mol
Melting Pt:250 °C (dec., lit.)
Density:2.338 g/mL (25 °C) (lit.)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Assay:≥99.0%
- pH (5% aq soln):4.1-4.5
Specification Test Results
Frequently Bought Together