Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
Fludrocortisone acetate
 
undefined
 
 
Fludrocortisone acetate

 

  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 % (105°C)
  • Solubility:
    (5 % in acetone) Clear colorless to light yellow
  • HPLC:
    ≥96.0 %

 

Frequently Bought Together