
Sarpogrelate hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:135159-51-2
:
Formula:C₂₄H₃₁NO₆·HCl
MDL Number:MFCD00887582
:465.97 g/mol
Melting Pt:145…148 °C
Storage Temperature:Refrigerator
UN:3077
ADR:9,III
:GHS09
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:146 to 152 °C
- Water:≤0.5 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together