Order Entry
Czech Republic
ContactUsLinkComponent
Ceftazidime pentahydrate 98%
Health
Ceftazidime pentahydrate 98%
Certificates
undefined
Health
Ceftazidime pentahydrate 98%

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    13 to 15 % (0.1g)
  • HPLC:
    ≥97.5 %

Specification Test Results

Frequently Bought Together