Mannose triflate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:92051-23-5
:
Formula:C₁₅H₁₉F₃O₁₂S
MDL Number:MFCD00012353
:480.36 g/mol
Melting Pt:118…122 °C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:118 to 122 °C
- Solubility:Clear colorless to slightly yellow
- HPLC:≥97.5 % (ELSD)
- Specific optical rotation:-12° to -19° (20 °C, 589 nm) (1 % in CH3CN)
Specification Test Results
Frequently Bought Together