
Mefloquine hydrochloride
Catalog # ACRO461130010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥97.5 % (on dry substance)
- Water:≤3 %
- HPLC:≥97.5 %
Specification Test Results
About this item
:
:414.78 g/mol
Melting Pt:253 °C
:GHS07
Storage Temperature:Ambient
CAS Number:51773-92-3
Formula:C₁₇H₁₆F₆N₂O
MDL Number:MFCD00797519