Order Entry
Czech Republic
ContactUsLinkComponent
Gemcitabine hydrochloride 98%
Irritant
Health
Gemcitabine hydrochloride 98%
Certificates
undefined
Irritant
Health
Gemcitabine hydrochloride 98%

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    98 to 102 %
  • HPLC:
    ≥97.5 %
  • Optical rotation:
    +43° to +50° (1 % in water)

Specification Test Results

Frequently Bought Together