

Oxyclozanide 98%
Catalog # ACRO460881000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Cream Powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 %
- HPLC:≥97.5 %
Specification Test Results
About this item
:
:401.46 g/mol
Storage Temperature:Ambient
UN:3077
ADR:9,III
:GHS07
:GHS09
CAS Number:2277-92-1
Formula:C₁₃H₆Cl₅NO₃
MDL Number:MFCD00864507