Order Entry
Czech Republic
ContactUsLinkComponent
 
Methyl palmitoleate 99% analytical standard for GC
Catalog # ACRO460400010
CAS Number:  
undefined
Methyl palmitoleate 99% analytical standard for GC
Catalog # ACRO460400010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4500 to 1.4520 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

About this item

 :
 :268.44 g/mol
Melting Pt:.5 °C
Density:0.88 g/cm³
Storage Temperature:Freezer
CAS Number:1120-25-8
Formula:C₁₇H₃₂O₂
MDL Number:MFCD00042911