

Methyl-4-fluorobenzoate 98%
:403-33-8
:
:C₈H₇FO₂
:MFCD00017959
:154.14 g/mol
:90…92 °C (27hPa)
:1.192 g/cm³
:78 °C
:GHS05
:GHS07
:Ambient
- Appearance:Colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4930 to 1.4950 (20°C, 589 nm)
- GC:≥97.5 %
Frequently Bought Together