Order Entry
Czech Republic
ContactUsLinkComponent
cis,cis-1,3,5-Cyclohexanetriol dihydrate 98%
cis,cis-1,3,5-Cyclohexanetriol dihydrate 98%
Certificates
undefined
cis,cis-1,3,5-Cyclohexanetriol dihydrate 98%

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Water:
    19 to 24 %
  • GC:
    ≥97.5 %

Specification Test Results

Frequently Bought Together