Order Entry
Czech Republic
ContactUsLinkComponent
Amorolfine hydrochloride 98%
Irritant
Environment
Amorolfine hydrochloride 98%
Certificates
undefined
Irritant
Environment
Amorolfine hydrochloride 98%

Specification Test Results

  • Appearance:
    White to light yellow to beige Crystalline powder or powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    206 to 211 °C
  • HPLC:
    ≥98.5 %

Specification Test Results

Frequently Bought Together