2'-Deoxycytidine hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3992-42-5
:
Formula:C₉H₁₄ClN₃O₄
MDL Number:MFCD00012840
:263.68 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:Off-white to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Water:≤8 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together