
Donepezil hydrochloride
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:120011-70-3
:
Formula:C₂₄H₂₉NO₃
MDL Number:MFCD00881312
:415.96 g/mol
Melting Pt:211…212 °C
UN:2811
:GHS06
Storage Temperature:Freezer
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Melting point:223 to 233 °C
- Water:≤0.5 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together