Order Entry
Czech Republic
ContactUsLinkComponent
2-Fluoro-4-hydroxybenzoic acid 98%
Irritant
2-Fluoro-4-hydroxybenzoic acid 98%
Catalog # ACRO457590250
CAS Number:  
undefined
Irritant
2-Fluoro-4-hydroxybenzoic acid 98%
Catalog # ACRO457590250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to light brown Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :156.1112 g/mol
Melting Pt:187…188 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:65145-13-3
Formula:C₇H₅FO₃
MDL Number:MFCD01310985