
2-Methylpyrrolidine hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:54677-53-1
:
Formula:C₅H₁₁N
MDL Number:MFCD08274961
:121.61 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:68 to 77 °C
- Titration with HClO4:≥96.0 %
Specification Test Results
Frequently Bought Together