
Tetrahydropyran-4-carboxylic acid 97%
Catalog # ACRO456720050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to yellow to brown Crystalline powder or solid
- Infrared spectrum:Conforms
- Melting point:87 to 91 °C
- Water:≤1.5 %
- Titration with NaOH:96.5 to 103.5 %
Specification Test Results
About this item
:
:130.14 g/mol
Boiling Pt:115 °C (20 mmHg)
Melting Pt:87…89 °C
:GHS07
Storage Temperature:Ambient
CAS Number:5337-03-1
Formula:C₆H₁₀O₃
MDL Number:MFCD00031016