
1-Phenyl-2-propyn-1-one 97%
Catalog # ACRO456430010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:1 g
- Sekvence:3047
Specifications
Specification Test Results
- Appearance:White to yellow Powder or crystalline powder
- Infrared spectrum:Conforms
- Melting point:48 to 53 °C
- GC:≥96.0 %
Specification Test Results
About this item
:
:130.15 g/mol
Boiling Pt:90 °C (,8 mmHg)
Melting Pt:51…52 °C
:GHS07
Storage Temperature:Ambient
CAS Number:3623-15-2
Formula:C₉H₆O
MDL Number:MFCD06661646