Order Entry
Czech Republic
ContactUsLinkComponent
1,4-Diazabicyclo[2.2.2]octanebis(sulphur dioxide)adduct 97%
Flammable
Irritant
1,4-Diazabicyclo[2.2.2]octanebis(sulphur dioxide)adduct 97%
Certificates
undefined
Flammable
Irritant
1,4-Diazabicyclo[2.2.2]octanebis(sulphur dioxide)adduct 97%

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    168 to 172 °C
  • Assay:
    ≥96.0 %

Specification Test Results

Frequently Bought Together