Order Entry
Czech Republic
ContactUsLinkComponent
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid 98%
Irritant
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid 98%
Catalog # ACRO455870050
CAS Number:  
undefined
Irritant
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid 98%
Catalog # ACRO455870050
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :197.96 g/mol
Melting Pt:127…129 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:603122-84-5
Formula:C₈H₈BFO₄
MDL Number:MFCD08436034