
2-(Methylsulphonyl)phenol 98%
:27489-33-4
:
:C₇H₈O₃S
:MFCD00127516
:88…90 °C
:GHS07
:Refrigerator
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:87 to 92 °C
- HPLC:≥97.5 %
Frequently Bought Together