
4-Cyano-1-butyne 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19596-07-7
:
Formula:C₅H₅N
MDL Number:MFCD00041563
:79.999 g/mol
Boiling Pt:79 °C (39 mmHg)
Density:0.9063 g/cm³
Flash Pt:65.5 °C
Storage Temperature:Refrigerator
UN:3276
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4340 to 1.4380 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together