Nystatin 85+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1400-61-9
:
Formula:C₄₇H₇₅NO₁₇
MDL Number:MFCD25541651
:926.09 g/mol
Melting Pt:>160 °C
Storage Temperature:Freezer
Specification Test Results
- Appearance:Yellow to light brown Powder
- Infrared spectrum:Conforms
- Water:≤6 %
- HPLC:≥85.0 %
- UV:at 305 nm A: ≥0.60
- Activity:≥4400 IU/mg (dried substance)
Specification Test Results
Frequently Bought Together