

Fluconazole 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:86386-73-4
:
Formula:C₁₃H₁₂F₂N₆O
MDL Number:MFCD00274549
:306.28 g/mol
Melting Pt:137…141 °C
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:138 to 142 °C
- Loss on drying:≤1 % (105 °C, 3 h)
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together