Ethanolamine ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Aminoethanol, Monoethanolamine
CAS Number:141-43-5
:
Formula:NH₂CH₂CH₂OH
MDL Number:MFCD00008183
Boiling Pt:170 °C (760 mmHg)
Melting Pt:10.3 °C
Density:1.012 g/cm³
Molecular Weight:61.08 g/mol
Flash Pt:92 °C
Storage Temperature:Ambient
ADR:8 III
Merck Index:13,03762
UN:2491
:GHS07
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.30 %
- GC:≥99.0 %
- Color scale:≤15 APHA
- Iron (Fe):≤5 ppm
- Heavy metals (as Pb):≤5 ppm
Specification Test Results
Frequently Bought Together