
4-Fluoro-2-methoxypyridine 97%
Catalog # ACRO450010010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorlesss to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4740 to 1.4760 (20°C, 589 nm)
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:127.12 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:96530-81-3
Formula:C₆H₆FNO
MDL Number:MFCD09835267