Order Entry
Czech Republic
ContactUsLinkComponent
3-Fluoroazetidine hydrochloride 95%
Irritant
3-Fluoroazetidine hydrochloride 95%
Catalog # ACRO449990010
CAS Number:  
undefined
Irritant
3-Fluoroazetidine hydrochloride 95%
Catalog # ACRO449990010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to yellow Powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥94 %

Specification Test Results

About this item

 :
 :111.55 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:617718-46-4
Formula:C3H7ClFN
MDL Number:MFCD05663715