
6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid 97%
Katalogové číslo ACRO449910050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to brown Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 %
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:241.05 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:944896-42-8
Formula:C₈H₅BrN₂O₂
MDL Number:MFCD10698063