Order Entry
Czech Republic
ContactUsLinkComponent
n-Hexane 95+% ACS
Irritant
Health
Environment
Flammable
n-Hexane 95+% ACS
Certificates
undefined
Irritant
Health
Environment
Flammable
n-Hexane 95+% ACS

Specification Test Results

  • Appearance:
    Clear liquid
  • GC:
    ≥95.0 % (n-Hexane)
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.001 %
  • Sulfur compounds (as S):
    ≤0.005 %
  • Water soluble titr. acid:
    ≤0.0003 meg/g
  • Thiophene:
    Passes test

Specification Test Results

Related Information

Frequently Bought Together