Order Entry
Czech Republic
ContactUsLinkComponent
Rifampicin 95%
Rifampicin 95%
Certificates
undefined
Rifampicin 95%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Light orange to red-brown Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤2 % (100 mg, 60°C, 4 h) (vacuum)
  • HPLC:
    ≥94 %

Specification Test Results

Frequently Bought Together