
4-Aminopyridine-2-carboxylic acid 97%
Catalog # ACRO455772500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:Beige Powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:138.13 g/mol
Melting Pt:319 °C
:GHS07
Storage Temperature:Ambient
CAS Number:100047-36-7
Formula:C₆H₆N₂O₂
MDL Number:MFCD02685671