
N-Benzylhydroxylamine hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:29601-98-7
:
Formula:C₇H₁₀ClNO
MDL Number:MFCD00043377
:159.6134 g/mol
Melting Pt:108…110 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder or crystals
- Infrared spectrum:Conforms
- Melting point:108 to 112 °C
- Titration Argentometric:97.5 to 102.5 %
Specification Test Results
Frequently Bought Together