



Lithiumbis(trimethylsilyl)amide 1 M in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4039-32-1
:
Formula:C₆H₁₈LiNSi₂
MDL Number:MFCD00008261
:167.33 g/mol
Boiling Pt:65 °C (760 mmHg)
Density:0.9 g/cm³
Flash Pt:–21 °C
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light yellow to amber Liquid
- Strength:0.90 to 1.10 M
Specification Test Results
Related Information
Frequently Bought Together