Reagent/centrifuge tubes, flat base, Sarstedt
DESG62.470
: DESAGA
- Closure type:Without cap
- Ø×L:16,5×95 mm
- Packed:1000/case
- Type:with writing area
- Capacity:12 ml
- Environmentally Preferable:
- Pk:1.000
- RCF max.:3500×g
PS, clear, for push-in stoppers.
- With writing space
- With graduation
- Non sterile
Universal tubes ideal for a wide range of applications.
: Stoppers must be ordered seperately.