VWR®, Rotors for Ministar Centrifuge
Supplier: VWR Collection
Certificates
About this item
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Decontamination solution, DNA AWAY®
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Disposable weighing dishes
Supplier: LUDI
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
Bags, zip seal, writing area, Minigrip® bio-based
Supplier: Flexico