DNA ladder, low range, MassRuler
Catalog # SM0383
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 KIT
- Description:MassRuler low range DNA ladder, ready-to-use, 80 - 1031 bp, 2×500 µl
Specifications
About this item
MassRuler Low Range DNA Ladder, ready to use, 80 to 1031 bp, is specially designed for accurate quantification and sizing of DNA fragments by agarose gel electrophoresis.
- Sharp bands
- Easy to remember fragment sizes and quantities
- Premixed with 6X MassRuler DNA Loading Dye for direct loading and room temperature storage
The intensity of the fragment in each ladder is calibrated against a standard that guarantees the precise quantity of each band. The ladders are mixtures of chromatography-purified, individual DNA fragments.
Delivery information: Supplied with 1 ml 6X loading dye for sample DNA.