Fluconazole, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
4 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1271700.
- 1271711.
- 1271722.
- 1271733.
- Description:
- Fluconazole
- Fluconazole Related Compound A
- Fluconazole Related Compound B
- Fluconazole Related Compound C
- Synonym(s):
- 2-[2-fluoro-4-(1H-1,2,4-triazol-1-yl)phenyl]-1,3-bis(1H-1,2,4-triazol-1-yl)-propan-2-ol
- 2-(4-fluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)-propan-2-ol
- 1,1'-(1,3-phenylene)di(1H-1,2,4-triazole)
- CAS No.:
- 86386-73-4
- 871550-15-1
- 81886-51-3
- 514222-44-7
- Formula:
- C₁₃H₁₂F₂N₆O
- C₁₃H₁₃FN₆O
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1271700.FluconazoleSpecifications:
More
Specifications
Cat. No.1271700.DescriptionFluconazoleSynonym(s)CAS No.86386-73-4FormulaC₁₃H₁₂F₂N₆O - 1271711.Fluconazole Related Compound AFluconazole Related Compound AUSP reference standardVial10 mgLog in for availability1 * 10 mg27 820,00 KčSpecifications:
More
Specifications
Cat. No.1271711.DescriptionFluconazole Related Compound ASynonym(s)2-[2-fluoro-4-(1H-1,2,4-triazol-1-yl)phenyl]-1,3-bis(1H-1,2,4-triazol-1-yl)-propan-2-olCAS No.871550-15-1Formula - 1271722.Fluconazole Related Compound BFluconazole Related Compound BUSP reference standardVial10 mgLog in for availability1 * 10 mg26 340,00 KčSpecifications:
More
Specifications
Cat. No.1271722.DescriptionFluconazole Related Compound BSynonym(s)2-(4-fluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)-propan-2-olCAS No.81886-51-3FormulaC₁₃H₁₃FN₆O - 1271733.Fluconazole Related Compound CFluconazole Related Compound CUSP reference standardVial10 mgLog in for availability1 * 10 mg25 820,00 KčSpecifications:
More
Specifications
Cat. No.1271733.DescriptionFluconazole Related Compound CSynonym(s)1,1'-(1,3-phenylene)di(1H-1,2,4-triazole)CAS No.514222-44-7Formula
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



