

Morpholin-3-one 98%
:109-11-5
:
:C₄H₇NO₂
:MFCD00631009
:101.1039 g/mol
:106…108 °C
:GHS05
:GHS07
:Ambient
- Appearance:Colorless to white Crystalline solid
- Infrared spectrum:Conforms
- Melting point:102 to 109 °C
- GC:≥97.5 %
Frequently Bought Together