Order Entry
Czech Republic
ContactUsLinkComponent
Methyl myristate 99% analytical standard for GC
Methyl myristate 99% analytical standard for GC
Certificates
undefined
Methyl myristate 99% analytical standard for GC
Certificates

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4360 to 1.4380 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together