Boron nitride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10043-11-5
:
Formula:BN
MDL Number:MFCD00011317
:24.82 g/mol
Melting Pt:2700…3000 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder
- Assay:≥97.5 %
- Particle size:0.17 to 5 µm
Specification Test Results
Frequently Bought Together