Test tube rack slanter, SP Bel-Art
Katalogové číslo BELAF188900100
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Rack slanter
- Pk:1
Specifications
About this item
Made of PP.
- Base legs have predetermined slots that stand slides in and out of to easily change angles
- Fits racks up to 293×242×83 mm
- Autoclavable
These rack slanters used to tilt the test tube rack to 25°, 35° or 45° to reduce hand strain when working with pipettors. It is also ideal for creating agar slant tubes.