Wire shelves
Wire shelf, fits shelf length 568 mm, Stainless steel, 2,7 kg
Katalogové číslo METOLT30WS
Dodavatel: Metro International
Přejít na:
Product Details & Documents
Stainless steel wire shelf mount on 3 sided frame with use of clips.
Specifikace
- Description:Wire shelf, fits shelf length 568 mm
- Material:Stainless steel
- Weight:2,7 kg



